Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Admittance Signs: DANGER - EXPLOSIVES KEEP OUT
Identify equipment, rooms and materials which contain or are contaminated with hazardous agents
$36.75 - $93.00
Specifications
Quantity | 1 Ea. |
---|---|
Mounting Type | Surface Mount |
For Use With (Application) | Designed for use in industrial, utility, commercial and institutional environments |
Height (English) | 10 in. |
Height (Metric) | 25.40 cm |
Catalog Number | Mfr. No. | Material | Temperature Range (English) | Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Temperature Range (English) | Thickness | Price | Quantity & Availability | |||
19-035-950
|
Brady™
25659 |
Plastic | -40°F to 175°F | 0.06 in. |
Each for $36.75
|
|
|||
19-035-948
|
Brady™
70330 |
Fiberglass | -40°F to 193°F | 0.1 in. |
Each for $93.00
|
|
|||
19-035-952
|
Brady™
85174 |
Polyester | -40°F to 230°F | 0.006 to 0.01 in. |
Each for $67.00
|
|
|||
Description
- Available in three materials - plastic, fiberglass and polyester
- Black/red on white
Specifications
1 Ea. | |
Designed for use in industrial, utility, commercial and institutional environments | |
25.40 cm | |
14 in. | |
Explosive Keep Out |
Surface Mount | |
10 in. | |
-40°F | |
35.56 cm | |
Admittance Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title