Learn More
Ampco Safety Tools Box End Wrench (Metric)
AMPCO™ Box End Wrench performs in high-risk and demanding environments.
$197.00 - $2712.00
Specifications
Product Type | Box End Wrench |
---|---|
Material | Aluminum Bronze |
Certifications/Compliance | Factory Mutual (FM) Approved |
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog Number | Mfr. No. | Drive Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Drive Size | Price | Quantity & Availability | |||||
19-173-403
|
Ampco Safety Tools
4400 |
14 mm |
Each for $197.00
|
|
|||||
19-173-404
|
Ampco Safety Tools
4402 |
15 mm |
Each for $206.00
|
|
|||||
19-173-405
|
Ampco Safety Tools
4404 |
16 mm |
Each for $216.00
|
|
|||||
19-173-406
|
Ampco Safety Tools
4406 |
17 mm |
Each for $216.00
|
|
|||||
19-173-407
|
Ampco Safety Tools
4408 |
18 mm |
Each for $270.00
|
|
|||||
19-173-408
|
Ampco Safety Tools
4410 |
19 mm |
Each for $270.00
|
|
|||||
19-173-409
|
Ampco Safety Tools
4412 |
20 mm |
Each for $270.00
|
|
|||||
19-173-410
|
Ampco Safety Tools
4414 |
21 mm |
Each for $306.00
|
|
|||||
19-173-411
|
Ampco Safety Tools
4416 |
22 mm |
Each for $306.00
|
|
|||||
19-173-412
|
Ampco Safety Tools
4418 |
23 mm |
Each for $306.00
|
|
|||||
19-173-413
|
Ampco Safety Tools
4420 |
24 mm |
Each for $361.00
|
|
|||||
19-173-414
|
Ampco Safety Tools
4422 |
25 mm |
Each for $361.00
|
|
|||||
19-173-415
|
Ampco Safety Tools
4424 |
26 mm |
Each for $361.00
|
|
|||||
19-173-416
|
Ampco Safety Tools
4426 |
27 mm |
Each for $396.00
|
|
|||||
19-173-417
|
Ampco Safety Tools
4428 |
28 mm |
Each for $396.00
|
|
|||||
19-173-418
|
Ampco Safety Tools
4430 |
29 mm |
Each for $396.00
|
|
|||||
19-173-419
|
Ampco Safety Tools
4432 |
30 mm |
Each for $408.00
|
|
|||||
19-173-420
|
Ampco Safety Tools
4434 |
32 mm |
Each for $443.00
|
|
|||||
19-173-421
|
Ampco Safety Tools
4436 |
33 mm |
Each for $443.00
|
|
|||||
19-173-422
|
Ampco Safety Tools
4438 |
34 mm |
Each for $499.00
|
|
|||||
19-173-423
|
Ampco Safety Tools
4440 |
35 mm |
Each for $499.00
|
|
|||||
19-173-424
|
Ampco Safety Tools
4442 |
36 mm |
Each for $499.00
|
|
|||||
19-173-425
|
Ampco Safety Tools
4444 |
37 mm |
Each for $499.00
|
|
|||||
19-173-426
|
Ampco Safety Tools
4446 |
38 mm |
Each for $499.00
|
|
|||||
19-173-427
|
Ampco Safety Tools
4448 |
41 mm |
Each for $618.00
|
|
|||||
19-173-428
|
Ampco Safety Tools
4450 |
42 mm |
Each for $618.00
|
|
|||||
19-173-429
|
Ampco Safety Tools
4452 |
46 mm |
Each for $721.00
|
|
|||||
19-173-430
|
Ampco Safety Tools
4454 |
47 mm |
Each for $721.00
|
|
|||||
19-173-431
|
Ampco Safety Tools
4456 |
50 mm |
Each for $915.00
|
|
|||||
19-173-432
|
Ampco Safety Tools
4458 |
52 mm |
Each for $915.00
|
|
|||||
19-173-433
|
Ampco Safety Tools
4460 |
54 mm |
Each for $915.00
|
|
|||||
19-173-434
|
Ampco Safety Tools
4462 |
55 mm |
Each for $1,220.00
|
|
|||||
19-173-435
|
Ampco Safety Tools
4464 |
56 mm |
Each for $1,220.00
|
|
|||||
19-173-436
|
Ampco Safety Tools
4466 |
58 mm |
Each for $1,220.00
|
|
|||||
19-173-437
|
Ampco Safety Tools
4468 |
60 mm |
Each for $1,528.00
|
|
|||||
19-173-438
|
Ampco Safety Tools
4470 |
63 mm |
Each for $1,528.00
|
|
|||||
19-173-439
|
Ampco Safety Tools
4472 |
65 mm |
Each for $1,710.00
|
|
|||||
19-173-440
|
Ampco Safety Tools
4474 |
67 mm |
Each for $1,710.00
|
|
|||||
19-173-441
|
Ampco Safety Tools
4476 |
70 mm |
Each for $1,935.00
|
|
|||||
19-173-442
|
Ampco Safety Tools
4478 |
71 mm |
Each for $1,935.00
|
|
|||||
19-173-443
|
Ampco Safety Tools
4480 |
75 mm |
Each for $2,184.00
|
|
|||||
19-173-444
|
Ampco Safety Tools
4482 |
77 mm |
Each for $2,184.00
|
|
|||||
19-173-445
|
Ampco Safety Tools
4484 |
80 mm |
Each for $2,712.00
|
|
|||||
19-173-446
|
Ampco Safety Tools
4486 |
82 mm |
Each for $2,712.00
|
|
|||||
Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Box End Wrench | |
Aluminum Bronze | |
Factory Mutual (FM) Approved |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.