Learn More
Biosys Laboratories Biosys UriScan™ Urine Test Strips
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For measuring different parameters in urine sample
$36.25 - $485.00
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
23-679-104
![]() |
Biosys Laboratories
U20 |
2 GP strips for measuring glucose, protein |
Pack of 100 for $36.25
Case of 10 PK for $362.00
|
|
|||||
23-679-103
![]() |
Biosys Laboratories
U25 |
6L strips for measuring blood, protein, nitrite, pH, glucose, leucocytes |
Case of 1000 for $485.00
|
|
|||||
23-679-100
![]() |
Biosys Laboratories
U39 |
10SGL strips for measuring blood, bilirubin, urobilinogen, ketones, protein, nitrite, glucose, pH, specific gravity, leucocytes |
Pack of 100 for $59.00
Case of 10 PK for $589.00
|
![]() |
|||||
Specifications
UriScan™ Urine Test Strips |