Learn More
ILC Dover BioShield™ Full Hood
Click to view available options
Quantity:
25/Pk.
3/Pk.
Description
- Visor is impact resistant, made with optical grade polycarbonate
- Internal and external skirt interfaces with most protective suits and requires no fit testing
Specifications
Specifications
Material | Polycarbonate, Polyolefin |
Latex Free | Yes |
Quantity | 25/Pk. |