Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide CN HPLC Column, 160 Å, 2.7 μm Particle Size
BIOshell™ 160 Å Peptide CN columns provide an alternative selectivity to the traditional C18 bonded phases.
Supplier: MilliporeSigma™ Supelco™ 66966U
Description
The introduction of the cyano group enables peptides and small proteins to be separated due to a combination of hydrophobicity and dipole-dipole interactions making this column an excellent choice for biomolecules that are not resolved on traditional C18 columns. These columns contain advanced Fused-Core particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping. Additionally, the sterically-protected cyanopropyl ligands provide extra stability allowing the columns to be used at an extended pH range (2-9) and high temperatures (up to 90°C).Specifications
2.1 mm | |
2.7 μm | |
160 Å | |
BIOshell | |
1.5% | |
Reversed Phase | |
Fused-Core HPLC |
5 cm | |
1 to 8 | |
Cyano | |
Cyano Phase/Spherical Silica Particle Platform | |
Yes | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BIOshell is a trademark of Sigma-Aldrich Co. LLC
Spot an opportunity for improvement?
Product Content Correction