Learn More
Electron Microscopy Sciences Micromesh™ Biopsy Cassettes in QuickLoad™ Stacks, Yellow
Supplier: Electron Microscopy Sciences 70079110
These Micromesh biopsy cassettes are compatible with Leica and Sakura printers, With 1676 square openings (0.38 mm) there is greatly improved fluid exchange without having to use biopsy pads. Large anterior and posterior slots in cassette and cover ensure the biopsy cassette will sink rapidly. A large square compartment with a side measuring 27 mm is perfect, even for needle biopsies. Molded from a special high density polymer, these patented cassettes keep specimens submerged and are resistant to the chemical action of histological solvents. The Micromesh mesh ensures efficient fluid exchange and drainage. The one-piece integral lid eliminates the need for separate steel lids. They can be opened and closed as often as necessary and relock securely without danger of specimen loss. The cover does not protrude above the cassette. Anterior writing area is at a 45 degree angle to make the cassette more suitable for automated cassette printers.Color: Yellow
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.