Learn More
Bioassay Systems EnzyFluo™ Bile Acid Assay Kit. For quantitative determination of total bile acids and evaluation of drug effects on bile acid metabolism.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems EFBA100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative determination of total bile acids and evaluation of drug effects on bile acid metabolism. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. Linear detection range of 1 - 150 µM bile acids. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL530/585nm. Samples: Serum, plasma, urine and other biological samples. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 1 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.