Learn More
BioMed Diagnostics Contains a BHBCheck Plus meter, a 25-count vial of blood ketone strips, a case for the readers and vials, and a user manual. | Detects: Blood ketone and glucose in bovine. | Available in: 12 case
Supplier: BioMed Diagnostics HD125
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The ultimate package! This starter kit contains a BHBCheck Plus meter, a 25-count vial of BHBCheck Plus Blood Ketone Strips, a hard carry case for the readers and vials, and a user manual. This is a great introductory item. | Detects: Blood ketone and glucose in bovine. | Available in: 12 case