Learn More
CTL Scientific Supply Corp BENZETHONIUM CHLORIDE 2.5L
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 30497.327
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Benzethonium chloride 0.004 mol/l AVS TITRINORM volumetric solution. Material used in quality control to check for purity in surfacants/detergents. 2.5 liter bottle