Learn More
Bel-Art™ SP Scienceware™ Polygon Spinbar™ Magnetic Stirring Bars
Adds turbulence relative to similar smooth size cylindrical bars.
$652.00 - $1605.00
Specifications
Certifications/Compliance FDA, USP Class VI, ISO 9001:2008
Color White
Material Alnico Magnet
Quantity 50
Shape Polygon
View More Specs
Products 8
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity & Availability  
03-410-522
View Documents
Bel-Art™
F371220012
0.17 in. 4.5 mm 0.47 in. 12 mm
Case of 50 for $652.00
Add to Cart
03-410-523
View Documents
Bel-Art™
F371220020
0.23 in. 6 mm 0.78 in. 20 mm
Case of 50 for $793.00
Edge
Add to Cart
03-410-524
View Documents
Bel-Art™
F371220025
0.23 in. 6 mm 0.98 in. 25 mm
Case of 50 for $819.00
Add to Cart
03-410-525
View Documents
Bel-Art™
F371220035
0.23 in. 6 mm 1.37 in. 35 mm
Case of 50 for $907.50
Add to Cart
03-410-526
View Documents
Bel-Art™
F371220040
0.31 in. 8 mm 1.57 in. 40 mm
Case of 50 for $869.00
Add to Cart
03-410-528
View Documents
Bel-Art™
F371220050
0.31 in. 8 mm 1.96 in. 50 mm
Case of 50 for $977.50
Add to Cart
03-410-529
View Documents
Bel-Art™
F371220060
0.39 in. 10 mm 2.36 in. 60 mm
Case of 50 for $1,103.00
Add to Cart
03-410-530
View Documents
Bel-Art™
F371220070
0.39 in. 10 mm 2.7 in. 70 mm
Case of 50 for $1,605.00
Add to Cart
Description
Description
Adds turbulence relative to similar smooth size cylindrical bars.
  • Marring of plastic containers diminished
  • Multifaceted surface adds turbulence relative to similar size smooth cylindrical stirring bars
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
Certifications
ISO 9001:2008
Specifications
Specifications
FDA, USP Class VI, ISO 9001:2008
Alnico Magnet
Polygon
White
50