Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) with Lecithin and Polysorbate 80 (LP80)
Specifications
Color | Light Beige |
---|---|
Manufacturing Origin | United States |
For Use With (Application) | Cultivation of fastidious microorganisms and visualization of hemolytic reactions |
Catalog Number | Mfr. No. | dimMediaType | Packaging Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimMediaType | Packaging Type | Quantity | Price | Quantity & Availability | |||
B215278
|
BD
215278 |
Single Wrapping, Aseptic Fill | 100/Pk. |
Pack of 100 for $628.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
B215276
|
BD
215276 |
Sterile Double Wrapping Plate | 100/Pk. |
Item Discontinued
Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. View Alternate Product |
N/A | ||||
B215275
|
BD
215275 |
Sterile Double Wrapping Plate | 10/Pk. |
Item Discontinued
Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. View Alternate Product |
N/A | ||||
B215285
|
BD
215285 |
VHP Resistant Triple Wrapping Isolator Pack | 100/Pk. |
Item Discontinued
Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. View Alternate Product |
N/A | ||||
Description
The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.
The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:
- Withstands an accidental drop from the height of a bench-top workspace
- Ensures the integrity of the sample
- Prevents the dish from opening unintentionally
- Protects media from accidental cross contamination
- Improves stability during stacking and storage
Specifications
Light Beige | |
United States | |
Cultivation of fastidious microorganisms and visualization of hemolytic reactions |
Safety and Handling
ShelfLife : 112 Days
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title