Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Basic Agarose Premier
Basic Agarose Premier, Analytical Grade
$577.00 - $1385.00
Specifications
CAS | 9012-36-6 |
---|---|
Description | Agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae. |
Color | White |
Product Type | Agarose |
Form | Powder |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP1AGAF0100
|
MP Biomedicals™
11AGAF0100 |
100 g |
Each for $577.00
|
|
|||||
MP1AGAF0500
|
MP Biomedicals™
11AGAF0500 |
500 g |
Each for $1,385.00
|
|
|||||
Description
- Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
- Basic agarose premier is ideal for routine rapid separation of DNA and RNA fragments as well as PCR products, the preparation of plasmids, and for screening, cloning and blotting techniques.
Specifications
9012-36-6 | |
White | |
Powder | |
Store at Room Temperature (15-30°C). |
Agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae. | |
Agarose | |
Analysis of nucleic acids bygel electrophoresis |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title