Learn More
LabScientific Bacti-Zapper, tube mouth, needle, and loop sterilizer, microsterilizer, adjustable tilt angle, no splatter, no open flame. 110V - 120V (USA & Canada)
Supplier: LabScientific BC2001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Sterilize loops and needles with infrared heat. Safeguards laboratory personnel. Prevents infections splatter and contamination. Adjustable tilt angle. Ideal for anaerobic chambers. Weighed base for stability. Operates on 110-120V 60Hz U.L. Approved. 220V and 240V Bacticinerators and Elements available for outside USA and Canada.