Learn More
Cole-Parmer Benchmark Scientific BactiZapper Infrared Sterilizer, 120 VAC
Supplier:  Cole-Parmer 0185022
Encompass_Preferred
The BactiZapper utilizes infrared heat to incinerate organic material deep within the ceramic funnel. Sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1500°F. Unit can be used in anaerobic chambers
Catalog No. NC1492119