Learn More
Andwin Scientific BACTERIA CELL SPREADER
Supplier: Andwin Scientific S66722
This bacteria cell spreader is constructed with stainless steel that provides sterilized spread. This product features a smooth triangular end. It is ideal for use with 9 cm plates. The spreading bar is 25 mm wide.