Learn More
Zymo Research Corporation XJb Autolysis Glycerol Stock, 1 ml 500x L-Arabinose, 1 ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Zymo Research Corporation T5041
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Additionally, it does not require use of any potentially interferingcomponents such as detergents, commonly found in various lytic buffers.Highlights• Simple and controlled autolysis of E. coli• Strains can by lysed in minutes after harvesting• Method compatible with most buffer systems• Ideal for protein expression and purification, also applicable for extraction of nucleic acids• Scale up to lyse more samples without increase in time or labor
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.