Learn More
Gurley Precision Instruments AUTO DENSOMETER/SMOOT TESTER
SDP
Supplier:  Gurley Precision Instruments 4340N
Encompass_Preferred
Request Bulk Order
Automatic Densometer & Smoothness TesterThe Model 4340 combines the abilities of our textile, standard and high pressure Densometers, S-P-S Testers as well as our programmable digital timers and provides a reading in seconds, thus reducing the time required to test a material. By utilizing state-of-the-art mass flow technology, the Model 4340 can accurately test both low and high flow materials traditionally tested between 0 and 50,000 'Gurley Seconds'. At the end of either an automatic or manual test, the user can then toggle between Gurley units, Bendtsen, Sheffield or Bekk equivalent units.
Catalog No. NC1865492