Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Audit™ MicroControls™ MicroDROP™ Urine Dipstick Control
SDP

Bi-level, human source materials fortified to target levels with compounds that produce the desired reaction when tested by the methods indicated

Supplier:  Audit™ MicroControls™ K065M4

Catalog No. 22-157-144

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Description

Description

  • Simulate human patient samples for monitoring precision of laboratory dipstick testing procedures for glucose, bilirubin, ketones, specific gravity, blood, pH; protein, urobilinogen, nitrite, hCG, leukocytes
  • Available in liquid form
  • Available sizes: 4 x 25mL

Specifications

Specifications

2°C to 8°C
DROP LQ Urine Dipstick Controls
2
Liquid
Acetest, Bilirubin Total (TBIL), Blood, Clinitest, Glucose, hCG, Ketones, Leukocytes, Microalbumin, Nitrite, pH, Total Protein, Specific Gravity, Urobilinogen
Testing Glucose, Bilirubin
4 x 25 mL
Safety and Handling

Safety and Handling

ShelfLife : 1 Month

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit