Learn More
Thermo Scientific ATC-HC ANION TRAP COLUMN
Supplier: Thermo Scientific 059604
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Ionpac ATC-HC AnionN Trap Column, 9 x 75mm, Packing Specification: Column: ATC-HC, Particle Diameter: 750micrometer, Substrate X-Linking: 8, Substrate Type: Microporous, Column Capacity mEq/column: 4.0, Functional Group: Quaternary Ammoni
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)