Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Worldwide™ Protective Products ATA™ M530 Cut-Resistant, High Dexterity Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Green
Navy
Orange
Description
- Uses ATA™ fiber technology to deliver a thin, form-fitting and lightweight design that supplies ANSI A4 cut protection appropriate for medium-to-high cut hazards
- The 13-gauge seamless knit gloves provide the dexterity and fingertip sensitivity you need to pick up broken glass, dissect samples, and work with task-based knives and scalpels
- Wear them under unlined nitrile gloves to add a cut resistant barrier
- The ATA™ fiber blend also uses Lycra fibers for lots of stretch, making gloves comfortable enough for extended periods of wear
- The Hide-Away technology provides long-lasting abrasion resistance
- Color coded hems make it easy to choose the right glove size
Specifications
Specifications
Coating Material | Uncoated |
Color | Green |
Cut Resistance Rating | ANSI A4 |
Hand | Specific |
Material | ATA Engineered Fiber |
Product Type | Cut-resistant Gloves |
Size | Large |
For Use With (Application) | Glass Handling, Metal Fabrication, Plastics and Molding, Use as a Liner Glove |
Glove Cuff Color | Brown |
Glove Cuff Style | Continuous Cuff |
Show More |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction