Learn More
Thermo Scientific ASSY,RING,SPRT 2/PK
Supplier:  Thermo Scientific 6040.0012
Encompass
Support ring for piston seals. 2 support rings. For use with UltiMate 3000 SD, RS pumps.
Catalog No. NC0425832
Explore available promotions