Learn More
Foxx Life Sciences The PUREGRIP Aspirator Bottles
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Foxx Life Sciences 1220038FLS
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The PUREGRIP Aspirator Bottles, featuring top thread and GL45 caps, have a tubular sidearm outlet near the bottom for attachment of flexible tubing and are very useful as a delivery/storage container for solutions. These bottles feature sturdy ring necks with a heavy duty rims. They can withstand repeated sterilization (wet or dry).
This item is not returnable.
View return policy