Learn More
Bioassay Systems EnzyChrom™ Aspartate Transaminase Assay Kit. For quantitative determination of aspartate transaminase (AST) activity and evaluation of drug effects on AST activity.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems EASTR100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative determination of aspartate transaminase (AST) activity and evaluation of drug effects on AST activity. Key Features: Sensitive. Linear detection range: 2-100 U/L. Simple and convenient. This simple, convenient assay can be carried out in a microplate or a cuvette and takes only 10 min. Method: OD340nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 2 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.