Learn More
Alkali Scientific ASI™ Charcoal Dextran Stripped Fetal Bovine Serum, 500mL Bottle
Supplier: Alkali Scientific FB125
Triple 0.1um sterile filtered, mycoplasma-tested and virus-screened, ships on dry ice. Store frozen at -20 to -10°C. Virus screened and mycoplasma tested. Lot number traceability.