Learn More
Showa Denko America Inc Asahipak NH2P-40 2E
Supplier: Showa Denko America Inc F7630010
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Suitable for saccharides analysis using HILIC mode, polyvinyl alcohol packing material provides excellent chemical stability and minimum deterioration, easily regenerated by washing in an alkaline solution, appropriate for evaporative light scattering detector (ELSD), corona charged aerosol detector (CAD), and LC/MS, USP L82 listed, dimensions: 2.0 mm ID x 250 mm L
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.