Learn More
Bioassay Systems QuantiChrom™ Arginase Assay Kit. For quantitative determination of arginase activity and screen for its inhibitors.
Supplier: Bioassay Systems DARG100
For quantitative determination of arginase activity and screen for its inhibitors. Key Features: Sensitive and accurate. Detection limit: 0.3 U/L for 2 hr arginase reaction in 96-well assay format. Simple and high-throughput. The procedure involves incubation of the provided substrate with the sample in a microplate followed by the addition of the coloring reagent. Can be readily automated as a high-throughput assay for thousands of samples per day. Method: OD430nm. Samples: Enzyme preparations, serum, plasma, tissue culture etc. Species: All. Procedure: Assay takes 2 hrs. Kit size: 100 tests. Detection limit: 1 U/L (0.3 U/L for 2 hr reaction).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.