Learn More
Felix Storch Inc 6 Cu.Ft. ADA Height Vaccine Refrigerator, Certified to NSF/ANSI 456 Vaccine Storage Standard
Supplier: Felix Storch Inc ARG61PVBIADA456
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
ARG61PVBIADA456 6CF GLASS DOOR, BUILT-IN, ADA HEIGHT IS CERTIFIED PURPSOE BUILT for SAFE VACCINE STORAGE. Accucold is proud to lead the industry with the largest line of cold storage product sizes certified to NSF/ANSI 456 Vaccine Storage Standard. The Pharma-Vac Performance Series supports meeting regulations and guidelines from the CDC, State Agencies, World Health Organization, and other Health Ministries on vaccine storage and handling. Temp. Security and Saftey Features: https://www.accucold.com/product/ARG61PVBIADA456
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.