Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Radians Arctic Radwear™ Cooling Wrap XT
Accelerates the evaporative cooling process
Supplier: Radians RCS20
Description
- For hot outdoor activities such as construction work, tough workouts or strenuous outdoor/indoor activity
- Soak for 2 to 3 min and wear for up to 5 hrs. of cooling relief (depending on humidity)
- Secure around neck by pulling through slot opening
- Reusable packaging design
- Safe and reusable
- Jobsite proven
- Anti-microbial treated to help prevent mold build-up and unwanted smells
- Heat stress safety management
- 100% polyester microfiber
Specifications
Polyester Microfiber | |
Blue | |
For hot outdoor activities such as construction work, tough workouts or any strenuous outdoor or indoor activity |
Cooling Wrap XT | |
32.5 x 12 in. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Warning: Cancer and reproductive harm
Spot an opportunity for improvement?
Provide Content Correction