Learn More
Thermo Scientific™ Arctic Express™ Dual Storage System PROMO
Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
$2273.60 - $2408.60
Specifications
Type Manual Fill LN2 Dewar
No. of Doors 1
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
CK50920PM
View Documents Promotion Details
Thermo Scientific™
CK50920PM
10 L
Each for $2,273.60
Add to Cart
CK50921PM
View Documents Promotion Details
Thermo Scientific™
CK50921PM
18.5 L
Each for $2,408.60
Add to Cart
Description
Description
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a “can and cane” storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
Specifications
Specifications
Manual Fill LN2 Dewar
1