Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anatomical Chart Series - The Muscular System Chart
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Shows anterior and posterior views of muscular system.
Supplier: Anatomical Chart Company 9781587790355
Description
- Classic illustrations by Peter Bachin
- Illustrates right half of the diaphragm, muscles of the posterior abdominal wall, and muscles of the right and right foot
- Size: 20W x 26 in. H
Specifications
Muscular System Anatomical Chart | |
Science | |
66.04 cm | |
50.8 cm |
Laminated | |
26 in. | |
20 in. | |
Anterior and posterior views of the whole body muscular system, right half of the diaphragm, muscles of the posterior abdominal wall, muscles of the right hand, muscles of the right foot, 2 eyelets for easy hanging |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction