Learn More
Enzo Life Sciences AMPIGENE 1-Step RT-PCR Kit (200 Reactions)
SDP
Supplier:  Enzo Life Sciences ENZKIT1050200
Encompass_Preferred
AMPIGENE 1-Step RT-PCR Kit uses the latest developments in reverse transcriptase (RTase) technology and buffer chemistry for efficient cDNA synthesis and PCR in a single tube. The modified MMLV RTase is blended with RNase inhibitor to prevent degradation of RNA by contaminating RNase. The RTase is thermostable and extremely active. An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. Applications: PCR.
Catalog No. 50-201-3810
Explore available promotions