Learn More
Hach Company Amino Acid Reagent Powder Pillows for LR Silica, 25 mL, pk/100
Supplier: Hach Company 80499
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For determination of low range silica by the Heteropoly Blue Method (0 - 2.0 mg/L SiO2). Optional reagent for high range Phosphate by the Amino Acid method. Sample Size: 25 mL. Pack of 100 Powder Pillows