Learn More
Ansell™ AlphaTec™ 56-910 Vinyl Closed-Back Aprons With Snap-on Ties
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- High-quality virgin vinyl in 8 mil thickness
- Medium-to-heavy-duty splash protection
- No exposed elastic or metal parts to create static and corrosion
- Permits cleaning to Class 100 standards
- Completely heat sealed and individually bagged
- Color: Blue
- Chemical processing, oil refining and petrochemicals
- Janitorial and maintenance work
- Beef/pork/poultry/fish processing
- Clean rooms
Order Info
Can be cleaned to ISO Class 5 (Class 100) cleanroom standards and shipped in heat-sealed bags; call your Fisher Safety Customer Service Representative at 1-800-772-6733 for details.
Compliance
Permeation tested in accordance with MIL-STD 282 (using various chemical warfare agents) and ASTM™ F739 (using ASTM F1001 test battery).
Specifications
Specifications
Size | 2X-Large |
Material | Vinyl |
Color | Blue |
Thickness | 8 mil |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.