Learn More
Ansell™ AlphaTec™ 56-007 PVC Aprons
Click to view available options
AlphaTec™ 56-007 PVC Aprons Medium Vinyl
Description
- Reliable Protection wherever oil, grease and chemicals are present
- Help workers keep clean and protected from harmful contact where splashes from acids and caustic bases are a concern
- Durable
- Offers resistance to abrasion and puncture
- Sealed grommets allow for size adjustment without tear-out
- Bib styles in four sizes with adjustable ties ensure optimal fit for a wide range of users
- Medium duty aprons feature 5-6 mil PVC and a sewn edge finish
- Uses:
- Food processing, Life sciences
Specifications
Specifications
Material | PVC, Vinyl |
Color | Blue |
Includes | 12/Pk., 6 bags/Cs. |
Dimensions (L x W) | 33 x 54 in. |
Style | AlphaTec 56-007 |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.