Learn More
Allegro™ Respirator Cleaning Kit
All the necessary components to clean and store full facepiece and half-mask respirators in easy-to-order kits.
$107.00 - $218.00
Specifications
Includes | Supplied with ninety 0.5 oz. (15 g) packets of water soluble cleaner, an air bulb, brush, 100 sterile storage bags with pressure sensitive seals that can be marked with cleaning information and a two sided 17 qt. (16 L) bucket |
---|---|
Certifications/Compliance | OSHA |
Includes: Supplied with 90 half-ounce (15g) packets of water-soluble cleaner, an air bulb, brush, 100 sterile storage bags with pressure-sensitive seals that can be marked with cleaning information, and a two-sided 17 qt. (16L) bucket.
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-088-600
|
Allegro™
400102 |
Replacement Soap |
Each for $107.00
|
|
|||||
19-087463
|
Allegro™
4001 |
Cleaning Kit |
Each for $205.00
|
|
|||||
19-088-526
|
Allegro™
4002 |
Cleaning Kit with Liquid Disinfectant Soap |
Each for $218.00
|
|
|||||
Description
Available with choice of a 32 oz. (0.9L) container of concentrated liquid disinfectant soap (EPA registered) or 90 half-ounce (15g) packets of cleaner.
Specifications
Supplied with ninety 0.5 oz. (15 g) packets of water soluble cleaner, an air bulb, brush, 100 sterile storage bags with pressure sensitive seals that can be marked with cleaning information and a two sided 17 qt. (16 L) bucket | |
OSHA |