Learn More
Newcomer Supply ALK-Lymphoma Cell Line Control Slides, 10/set
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Newcomer Supply 3018A
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Newcomer Supply ALK-Lymphoma (anaplastic lymphoma kinase) Cell Line Control Slides are for the positive immunohistochemical staining of ALK-Lymphoma with a NPM (nucleophosmin) gene translocation, associated with anaplastic large cell lymphoma (ALCL). The cell line control slides are developed from a process that allows production of compact cell preparations, cultured from human cell lines, that retain their cellular morphology and are tissue-like in composition. These cell lines are standardized and manufactured to provide consistent results from slide to slide. Each slide contains two, 2mm diameter cell line cores which demonstrate positive and negative expression for the specific marker. ALK-Lymphoma Control Slides contain sections of positive staining ALK-Lymphoma and negative staining breast adenocarcinoma.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.