Learn More
C. DIFF QUIK CHEK™
Rapid enzyme immunoassay for detecting glutamate dehydrogenase (GDH), also known as the C. difficile antigen, in stool specimens. Does not distinguish between toxigenic strains and those that cannot produce toxin. Developed and Manufactured by TECHLAB, Inc.
Supplier: Techlab Inc 30390
This item is not returnable.
View return policy
Description
- Test yields results in less than 30 minutes
- Samples may be performed in batches or individually for on-demand testing
- Works with any specimen consistency and with samples stored in transport media
Specifications
Moderate | |
C. difficile QUIK CHEK™ Test | |
25 Tests |
C. difficile GDH antigen | |
25 Membrane device pouches, diluent, wash buffer, conjugate, substrate, positive control, disposable plastic transfer pipettes |