Learn More
Chematics Inc ALCO'SCREEN 24/PK
Supplier: Chematics Inc 5500125
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Box of 24. All-Inclusive Alcohol Test Device. One-Step Alcohol Drug Test Device. For Forensic Use Only.For Criminal Justice Use Only. No Contact with Saliva Makes this Drug Test Tamper Proof. Approximates Blood Alcohol Content (BAC) from 0.02% to 0.30%. Uses Saliva to Test the Presence of Alcohol.