Learn More
Chematics Inc Alco-Screen 02 (Case of 288 Tests)
Supplier: Chematics Inc 56288
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Alco-Screen 02 is a simple, cost effective tool used to monitor alcohol consumption as part of a comprehensive Zero Tolerance testing program. Alco-Screen 02 is a Conforming Product for US Department of Transportation mandated testing of all transportation and safety sensitive employees for blood alcohol concentrations above the federally mandated zero tolerance level of 0.02%. Alco-Screen 02 is FDA 510(k) cleared for home use and is CLIA waived.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.