Learn More
Medchemexpress LLC HY-N0038 5mg Medchemexpress, Alantolactone CAS:546-43-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN00385MG
Encompass_Preferred
Medchemexpress, HY-N0038 5mg Alantolactone CAS:546-43-0 Alantolactone is a selective STAT3 inhibitor, with potent anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3365
Explore available promotions