Learn More
TARGETMOL CHEMICALS INC ALANTOLACTONE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2896
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alantolactone (Inula camphor)(Alant camphor) a sesquiterpene lactone has potential activity against triple-negative breast Y MDA-MB-231 cells by suppressing the signal transducer and activator of transcription 3 (STAT3) signaling pathway. purity: 99%
Catalog No. 50-258-0474
Explore available promotions