Learn More
CTL Scientific Supply Corp AG-FIX TEST STICKS,100 STRIPS
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CTL Scientific Supply Corp 90741
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
AG-FIX TEST STICKS - Test strip for the semi-quantitative determination of silver and the pH value in photographic solutions. 100 test strips . Gradation of scale - Silver: 0.5 - 1- 2 - 3 - 5 - 7 - 10 g/l Ag+. pH 4 - 5 - 6 - 7 - 8