Learn More
Agilent Technologies AffinityScript Multiple Temperature Reverse Transcriptase, 200 reactions per kit
Supplier:  Agilent Technologies 600109
Encompass
Our AffinityScript Multiple Temperature Reverse Transcriptase is engineered to be highly thermostable, allowing you to reverse transcribe at your preferred reaction temperature.
Catalog No. NC2032845
Explore available promotions