Learn More
Jade Scientific, Inc ACETONITRILE B&J HPLC 4L
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Jade Scientific, Inc BJAH0154/EA
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
acetonitrile UV, B&J Branda. Suggested application: HPLC, GC, Pesticide residue analysis, Spectrophotometry. Package Size: 20L. Container type is steel NOWPaK. CaS Number: 75-05-8. Hazard Class: 3. Package Group: II. Molecular Weight: 41.05. Density (g/mL) at 20a Deg. C: 0.7822. Boiling point (a Deg. C): 81.6. Freezing point (a Deg. C): -43.8. Flashpoint (a Deg. C): 6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.