Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PLTP Activity Assay Kit

To determine of the presence of PLTP, functional study

Supplier:  Abnova™ KA0791

Catalog No. 89-028-102


Add to Cart

Includes:
  • PLTP Donor molecule, 1mL
  • PLTP Acceptor molecule, 1mL
  • PLTP Assay buffer (10X), 5mL
  • Positive control (Rabbit Serum), 30μL

Description

Description

Specifications

Specifications

Store the kit at -80°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
Fluorometric
1mL PLTP Donor molecule, 1mL PLTP acceptor molecule, 5mL PLTP assay buffer 10X, 30μL positive control (Rabbit Serum)
Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine
PLTP Activity Assay Kit is used to determining of the presence of PLTP.
Functional Study
1 Kit
Activity Assay Kit
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit