Learn More
CUBE BIOTECH Nanodisc Copolymer AASTY 6-50
Supplier: CUBE BIOTECH 18262
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
10x1g of nanodisc copolymer AASTY 6-50 for the stabilization of membrane proteins outside the cell