Learn More
Kimberly-Clark™ Professional AA-9 WINDGARD Head Protection
Ideal for very cold environments
Supplier:  Kimberly-Clark™ Professional 16757
Encompass
Catalog No. 19-804-260
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Proban™ cotton inner shell maintains flame resistance through 25 industrial washes or 50 domestic washes
  • Nomex™ outer shell provides flame resistance throughout product life
  • Full head and neck liner with wide wraparound collar covers face and neck
  • Provides protection from electrical arc energy exposure
  • Leather/Nomex tabs attach to hard hats
  • One size
  • Color: Red
Product Suggestions
Product Suggestions