Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One 96-well Polystyrene Round Bottom Strip Plates, 1 x 8 strips
Suitable for agglutination tests. Greiner Bio-One 96 Well Polystyrene Round Bottom Strip Plates, 1 x 8 strips are suitable for non polar proteins and peptides.
Supplier: Greiner Bio-One 767070
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Pipetted simply and cleanly
- Suitable for +/-analyses
Specifications
Strip Plate | |
Medium Binding | |
Strip Plate | |
96 | |
Non-sterile | |
388 μL | |
Yes | |
Yes | |
Clear |
8 x 12 | |
White | |
Polystyrene | |
20 to 350 μL | |
Microlon 200 | |
Round Bottom (U-well) | |
100 | |
-20°C to +60°C |
Safety and Handling
ShelfLife : 4 Years after month of production
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction