Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 7502PF Nitrile Disposable Glove
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Designed for sensitive skin, the 7502PF nitrile disposable glove is powder, latex, and accelerator-free.
- The 2.5 mil nitrile provides good chemical protection from a wide array of chemical hazards.
- And featuring SHOWA's revolutionary Eco Best Technology (EBT), these gloves are engineered for accelerated biodegradation in biologically active landfills.
Specifications
Certifications/Compliance | ISO 374-1:2016, ISO 374-5:2016 |
Accelerator Free | Yes |
Applications | Laboratory |
Color | Blue |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | 2X-Large |
Show More |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction