Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6x-His Tag Monoclonal Antibody (4E3D10H2/E3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA1135
Description
By Western blot, MA1-135 detects both N-terminal and C-terminal His-tagged proteins.
6x-His Tag Monoclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
6x-His Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
4E3D10H2/E3 | |
Unconjugated | |
6His tag; 6X His; His Tag; Histidine Tag | |
Mixture of N-terminal and C-terminal 6x-His peptides, CG-HHHHHH and HHHHHH-GC | |
100 μg | |
Primary | |
Antibody | |
IgG1 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction